missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Chloro-2-fluoro-5-(trifluoromethyl)pyridine, 98%

Brand:  Alfa Aesar™ H64723.14

Additional Details : CAS Number : 72537-17-8 Weight : 0.02500kg Transport : UN number : 1993 Chem class : 3 Pack group : III

Quantity 25g

 View more versions of this product

Product Code. 15469106

  • 1357.04 DKK / 25g


Estimated Shipment date
from Supplier 18-02-2019
Add to basket

Specifications

Chemical Identifiers

72537-17-8
199.533
GDSROTVTTLUHCO-UHFFFAOYSA-N
1268075
C1=C(C=NC(=C1Cl)F)C(F)(F)F
C6H2ClF4N
MFCD00191918
3-chloro-2-fluoro-5-trifluoromethyl pyridine, 2-fluoro-3-chloro-5-trifluoromethyl pyridine, 2-fluoro-3-chloro-5-trifluoromethylpyridine, pyridine, 3-chloro-2-fluoro-5-trifluoromethyl, acmc-20aied, librarion l939, 3-chloro-2-fluoro-5 trifluoromethyl pyridine, 3-chloro-2-fluoro-5-trifluoromethyl-pyridine, 3-chloro-2-fluoro-5-trifluoromethyl-pyridine, 3-chloro-2-fluoro-5-trifluoromethyl pyridine
3-chloro-2-fluoro-5-(trifluoromethyl)pyridine

Specifications

3-Chloro-2-fluoro-5-(trifluoromethyl)pyridine
1.4330
Colorless
25g
C6H2ClF4N
UN1993
GDSROTVTTLUHCO-UHFFFAOYSA-N
3-chloro-2-fluoro-5-(trifluoromethyl)pyridine
1268075
Liquid
1.524g/mL
50°C to 55°C (11mmHg)
16°C to 20°C (decomposition)
72537-17-8
MFCD00191918
3-chloro-2-fluoro-5-trifluoromethyl pyridine, 2-fluoro-3-chloro-5-trifluoromethyl pyridine, 2-fluoro-3-chloro-5-trifluoromethylpyridine, pyridine, 3-chloro-2-fluoro-5-trifluoromethyl, acmc-20aied, librarion l939, 3-chloro-2-fluoro-5 trifluoromethyl pyridine, 3-chloro-2-fluoro-5-trifluoromethyl-pyridine, 3-chloro-2-fluoro-5-trifluoromethyl-pyridine, 3-chloro-2-fluoro-5-trifluoromethyl pyridine
C1=C(C=NC(=C1Cl)F)C(F)(F)F
199.533
199.53
98%
Videos
SDS