Få mere at vide
3-Chloro-2,4,6-trideuteroaniline, TRC
Artikelnummer. 30361042
Skift visning
Mængde:
500 mg
Artikelnummer. Mængde
30361042 500 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30361042 Leverandør Toronto Research Chemicals Leverandørnr. TRCC364612500MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Chloro-2,4,6-trideuteroaniline
CAS 347840-11-3
Molekylær formel C6 D3 H3 Cl N
Mængde 500 mg
Synonym 3-Chlorobenzenamine-d3, 3-Chlorobenzen-2,4,6-d3-amine, 3-Chloroaniline-2,4,6-D3, 3-Chlorobenzen-2,4,6-d3-amine (ACI), 3-Chloro-2,4,6-trideuterioaniline, 3-Chloro-2,4,6-trideuteroaniline, 3-Chloroaniline D3 (2,4,6-phenyl D3)
InChI formel InChI=1S/C6H6ClN/c7-5-2-1-3-6(8)4-5/h1-4H,8H2/i2D,3D,4D
SMIL [2H]c1cc([2H])c(Cl)c([2H])c1N
IUPAC navn 3-chloro-2,4,6-trideuterioaniline
Molekylvægt (g/mol) 130.59
Formel vægt 130.0377