Få mere at vide
3-Chloro-10,11-dihydro-5H-dibenzo[b,f]azepine, Mikromol™
Artikelnummer. 17912776
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17912776 100 mg
17972056 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17912776 Leverandør Mikromol Leverandørnr. MM0545.06
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3-Chloro-10,11-dihydro-5H-dibenzo[b,f]azepine
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Chloro-10,11-dihydro-5H-dibenzo[b,f]azepine
CAS 32943-25-2
Analyt- eller komponentnavne 3-Chloro-10,11-dihydro-5H-dibenzo[b,f]azepine
Forsendelsestilstand Room Temperature
Molekylær formel C14 H12 Cl N
Certificeringer/compliance ISO 9001
Synonym Clomipramine Hydrochloride Imp. F (EP), Clomipramine Imp. F (EP), Clomipramine USP Related Compound F, Clomipramine USP RC F, 3-Chloro-10,11-dihydro-5H-dibenzo[b,f]azepine, Clomipramine Related Compound F, Clomipramine Hydrochloride Impurity F, Clomipramine Impurity F
InChI formel InChI=1S/C14H12ClN/c15-12-8-7-11-6-5-10-3-1-2-4-13(10)16-14(11)9-12/h1-4,7-9,16H,5-6H2
SMIL Clc1ccc2CCc3ccccc3Nc2c1
IUPAC navn 2-chloro-6,11-dihydro-5H-benzo[b][1]benzazepine
Molekylvægt (g/mol) 229.7
Mængde 100 mg
Formel vægt 229.0658 g/mol