Få mere at vide
3-Chloro-1-(4-methylphenyl)propan-1-one, TRC
Artikelnummer. 30362872
Skift visning
Mængde:
5 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30362872 5 mg
30356596 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30362872 Leverandør Toronto Research Chemicals Leverandørnr. TRCC3688085MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Chloro-1-(4-methylphenyl)propan-1-one
CAS 22422-21-5
Molekylær formel C10 H11 Cl O
Mængde 5 mg
Synonym 3-Chloro-1-(4-methylphenyl)-1-propanone, 2-Chloroethyl 4-Methylphenyl Ketone, 3-Chloro-4'-methylpropiophenone, 4'-Methyl-3-chloropropiophenone, β-Chloroethyl p-tolyl Ketone
InChI formel InChI=1S/C10H11ClO/c1-8-2-4-9(5-3-8)10(12)6-7-11/h2-5H,6-7H2,1H3
SMIL Cc1ccc(cc1)C(=O)CCCl
IUPAC navn 3-chloro-1-(4-methylphenyl)propan-1-one
Molekylvægt (g/mol) 182.647
Formel vægt 182.05