Få mere at vide
(±)-3-Chloro-1,2-propane-d5-diol, CDN
Artikelnummer. 30322928
Skift visning
Mængde:
0.1 g
Pakningsstørrelse:
0.1g
Artikelnummer. Mængde unitSize
30322928 0.1 g 0.1g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322928 Leverandør CDN Leverandørnr. CDND18420.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Chloro-1,2-propanediol D5
CAS 342611-01-2
Molekylær formel C3 2H5 H2 Cl O2
Mængde 0.1 g
Synonym 3-MCPD D5, 3-chloro-1,2-propane-1,1,2,3,3-d5-diol
InChI formel InChI=1 S/C3H7ClO2/c4-1-3(6)2-5/h3,5-6 H,1-2H2/i1D2,2D2,3 D
SMIL [2 H]C([2 H])(O)C([2 H])(O)C([2 H])([2 H])Cl
IUPAC navn 3-chloro-1,1,2,3,3-pentadeuteriopropane-1,2-diol
Molekylvægt (g/mol) 115.57
Formel vægt 115.0448 g/mol
Procent renhed 98 atom % D, min 97% Chemical Purity