missing translation for 'onlineSavingsMsg'
Learn More

3-Carboxy-4-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™

Catalog Number 15477726
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
882679-10-9
C13H16BFO4
266.075
MFCD12546519
DXOXOBZCGBHKJS-UHFFFAOYSA-N
2-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid, 3-carboxy-4-fluorophenylboronic acid, pinacol ester, 3-carboxy-4-fluorophenylboronic acid,pinacol ester, 3-carboxy-4-fluorobenzeneboronic acid pinacol ester, 2-fluoro-5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid, benzoic acid, 2-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
46739598
2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)F)C(=O)O
This item is not returnable. View return policy
882679-10-9
C13H16BFO4
266.075
MFCD12546519
DXOXOBZCGBHKJS-UHFFFAOYSA-N
2-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid, 3-carboxy-4-fluorophenylboronic acid, pinacol ester, 3-carboxy-4-fluorophenylboronic acid,pinacol ester, 3-carboxy-4-fluorobenzeneboronic acid pinacol ester, 2-fluoro-5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoic acid, benzoic acid, 2-fluoro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
46739598
2-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(C=C2)F)C(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 266.07
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 3-Carboxy-4-fluorobenzeneboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'