Få mere at vide
3-Butenyl N,N-Bis(2-chloroethyl-d2)phosphorodiamidate, TRC
Artikelnummer. 30341042
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30341042 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30341042 Leverandør Toronto Research Chemicals Leverandørnr. TRCB69020710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Butenyl N,N-Bis(2-chloroethyl-d2)phosphorodiamidate
CAS 162377-56-2
Molekylær formel C8H13D4Cl2N2O2P
Mængde 10 mg
Synonym O-(3-Butenyl)-N,N-bis(2-chloroethyl-d2)phosphorodiamidate
InChI formel InChI=1S/C8H17Cl2N2O2P/c1-2-3-8-14-15(11,13)12(6-4-9)7-5-10/h2H,1,3-8H2,(H2,11,13)/i4D2,5D2
SMIL [2H]C([2H])(Cl)CN(CC([2H])([2H])Cl)P(=O)(N)OCCC=C
IUPAC navn N-[amino(but-3-enoxy)phosphoryl]-2-chloro-N-(2-chloro-2,2-dideuterioethyl)-2,2-dideuterioethanamine
Molekylvægt (g/mol) 279.14
Formel vægt 278.0656