Få mere at vide
3-Butenyl-d2 Tosylate, TRC
Artikelnummer. 30328852
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30328852 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30328852 Leverandør Toronto Research Chemicals Leverandørnr. TRCB755407100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Butenyl-d2 Tosylate
CAS 18932-24-6
Molekylær formel C11H12D2O3S
Mængde 100 mg
Synonym 1-Tosyloxy-3-butene-d2, 3-Buten-1-ol-d2 Tosylate, 3-Butenyl-d2 p-Toluenesulfonate, 3-(Butenyl-d2)-1-tosylate, But-3-enyl-d2 4-Methylbenzenesulfonate
InChI formel InChI=1S/C11H14O3S/c1-3-4-9-14-15(12,13)11-7-5-10(2)6-8-11/h3,5-8H,1,4,9H2,2H3/i9D2
SMIL [2H]C([2H])(CC=C)OS(=O)(=O)c1ccc(C)cc1
IUPAC navn 1,1-dideuteriobut-3-enyl 4-methylbenzenesulfonate
Molekylvægt (g/mol) 228.3
Formel vægt 228.0789