Få mere at vide
3-Bromotoluene-d7, CDN
Artikelnummer. 30310667
Skift visning
Mængde:
0.1 g
0.25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30310667 0.25 g
30317899 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30310667 Leverandør CDN Leverandørnr. CDND62870.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Bromotoluene-D7
CAS 1185318-69-7
Molekylær formel C7 D7 Br
Mængde 0.25 g
Synonym 1-Bromo-3-(trideuteriomethyl)-2,4,5,6-tetradeuteriobenzene, 1-Bromo-3-(methyl-d3)benzene-2,4,5,6-d4, 3-Bromotoluene-d7, 3-Bromotoluene-D7, 3-Bromotoluene-α,α,α,2,4,5,6-d7
InChI formel InChI=1 S/C7H7Br/c1-6-3-2-4-7(8)5-6/h2-5 H,1H3/i1D3,2 D,3 D,4 D,5 D
SMIL [2 H]c1c([2 H])c(Br)c([2 H])c(c1[2 H])C([2 H])([2 H])[2 H]
IUPAC navn 1-bromo-2,3,4,6-tetradeuterio-5-(trideuteriomethyl)benzene
Molekylvægt (g/mol) 178.0776
Formel vægt 177.0171 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity