Få mere at vide
3-Bromopyridine-d4, TRC
Artikelnummer. 30338869
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30338869 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30338869 Leverandør Toronto Research Chemicals Leverandørnr. TRCB68674250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Bromopyridine-d4
CAS 66148-14-9
Molekylær formel C5 D4 Br N
Mængde 50 mg
Synonym Pyridine-2,3,4,6-d4, 5-bromo- (9CI), 3-Bromopyridine-d4, 5-Bromopyridine-2,3,4,6-d4, 3-Bromopyridine-D4
InChI formel InChI=1S/C5H4BrN/c6-5-2-1-3-7-4-5/h1-4H/i1D,2D,3D,4D
SMIL [2H]c1nc([2H])c(Br)c([2H])c1[2H]
IUPAC navn 3-bromo-2,4,5,6-tetradeuteriopyridine
Molekylvægt (g/mol) 162.021
Formel vægt 160.978