missing translation for 'onlineSavingsMsg'
Learn More

3'-Bromoacetophenone, 97%, Thermo Scientific Chemicals

Catalog Number p-3735676
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2142-63-4
C8H7BrO
199.05
MFCD00000083
JYAQYXOVOHJRCS-UHFFFAOYSA-N
3'-bromoacetophenone, m-bromoacetophenone, 1-3-bromophenyl ethanone, 1-3-bromophenyl ethan-1-one, ethanone, 1-3-bromophenyl, 1-acetyl-3-bromobenzene, 3-bromoacetophenone, 1-3-bromo-phenyl-ethanone, 3-bromo acetophenone, acetophenone, 3'-bromo
16502
1-(3-bromophenyl)ethanone
CC(=O)C1=CC(=CC=C1)Br
This item is not returnable. View return policy
2142-63-4
C8H7BrO
199.05
MFCD00000083
JYAQYXOVOHJRCS-UHFFFAOYSA-N
3'-bromoacetophenone, m-bromoacetophenone, 1-3-bromophenyl ethanone, 1-3-bromophenyl ethan-1-one, ethanone, 1-3-bromophenyl, 1-acetyl-3-bromobenzene, 3-bromoacetophenone, 1-3-bromo-phenyl-ethanone, 3-bromo acetophenone, acetophenone, 3'-bromo
16502
1-(3-bromophenyl)ethanone
CC(=O)C1=CC(=CC=C1)Br

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Density 1.4900g/mL
Boiling Point 79.0°C to 81.0°C (2.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula BrC6H4COCH3
Refractive Index 1.5749 to 1.5769
Quantity 5 g
Specific Gravity 1.49
Formula Weight 199.05
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3'-Bromoacetophenone
Show More Show Less
compliance-icons
Product Identifier
  • 3'-Bromoacetophenone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'