missing translation for 'onlineSavingsMsg'
Få mere at vide

3-Bromo Nevirapine, TRC

Artikelnummer. 30336691
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30336691 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30336691 Leverandør Toronto Research Chemicals Leverandørnr. TRCB68598510MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 3-Bromo Nevirapine
CAS 284686-21-1
Molekylær formel C15H13BrN4O
Mængde 10 mg
Synonym 3-Bromo-11-cyclopropyl-5,11-dihydro-4-methyl-6H-dipyrido[3,2-b:2',3'-e][1,4]diazepin-6-one
InChI formel InChI=1S/C15H13BrN4O/c1-8-11(16)7-18-14-12(8)19-15(21)10-3-2-6-17-13(10)20(14)9-4-5-9/h2-3,6-7,9H,4-5H2,1H3,(H,19,21)
SMIL Cc1c(Br)cnc2N(C3CC3)c4ncccc4C(=Nc12)O
IUPAC navn 6-bromo-2-cyclopropyl-7-methyl-2,4,9,15-tetrazatricyclo[9.4.0.03,8]pentadeca-1(11),3,5,7,12,14-hexaen-10-one
Molekylvægt (g/mol) 345.19
Formel vægt 344.0273
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.