Learn More
Alfa Aesar™ 3-Bromo-N-(4-n-butylphenyl)-5-(trifluoromethyl)benzenesulfonamide, 97%
Brand:  Alfa Aesar™ H64863.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15469206
  • 1379.41 DKK / 1g
Estimated Shipment date
from Supplier 20-02-2019
Add to basket
Specifications
Chemical Identifiers
MFCD27665319
Specifications
3-Bromo-N-(4-n-butylphenyl)-5-(trifluoromethyl)benzenesulfonamide
MFCD27665319
Solid
1g
436.29
97%