missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-N-(3-fluorophenyl)-5-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64680.06

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15469066

  • 6330.39 DKK / 5g


Estimated Shipment date
from Supplier 20-02-2019
Add to basket

Specifications

Chemical Identifiers

MFCD27665303

Specifications

3-Bromo-N-(3-fluorophenyl)-5-(trifluoromethyl)benzenesulfonamide
MFCD27665303
Solid
5g
398.17
97%
Videos
SDS