missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromo-N-(2-fluoro-5-methylphenyl)-5-(trifluoromethyl)benzenesulfonamide, 97%

Brand:  Alfa Aesar™ H64117.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15408716

  • 1495.88 DKK / 1g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

3-Bromo-N-(2-fluoro-5-methylphenyl)-5-(trifluoromethyl)benzenesulfonamide
412.2
MFCD27665253

Specifications

3-Bromo-N-(2-fluoro-5-methylphenyl)-5-(trifluoromethyl)benzenesulfonamide
97%
412.2
Solid
1g
MFCD27665253
Videos
SDS