missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-pyrrolidinophenylboronic Acid, TRC

Product Code. 30344683
Change view
Click to view available options
Mængde:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30344683 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30344683 Supplier Toronto Research Chemicals Supplier No. TRCB695403250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 3-Bromo-5-pyrrolidinophenylboronic Acid
CAS 1256355-16-4
Molekylær formel C10H13BBrNO2
Mængde 250 mg
Synonym [3-Bromo-5-(pyrrolidin-1-yl)phenyl]boronic Acid, B-[3-Bromo-5-(1-pyrrolidinyl)phenyl]boronic Acid, 3-Bromo-5-(pyrrolidin-1-yl)phenylboronic Acid
InChI formel InChI=1S/C10H13BBrNO2/c12-9-5-8(11(14)15)6-10(7-9)13-3-1-2-4-13/h5-7,14-15H,1-4H2
SMIL OB(O)c1cc(Br)cc(c1)N2CCCC2
IUPAC navn (3-bromo-5-pyrrolidin-1-ylphenyl)boronic acid
Molekylvægt (g/mol) 269.93
Formel vægt 269.0223
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.