missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-iodopyridine, 95%, Thermo Scientific Chemicals

Catalog Number p-7055429
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
233770-01-9
C5H3BrIN
283.894
MFCD03086019
AOOZLVWDZUPEHT-UHFFFAOYSA-N
820409
3-bromo-5-iodopyridine
C1=C(C=NC=C1I)Br
This item is not returnable. View return policy
233770-01-9
C5H3BrIN
283.894
MFCD03086019
AOOZLVWDZUPEHT-UHFFFAOYSA-N
820409
3-bromo-5-iodopyridine
C1=C(C=NC=C1I)Br

Reactant in the synthesis chiral 4, 4?-Bipyridines. 3-bromo-5-(trifluoromethyl)pyridine is prepared from 3-Bromo-5-iodopyridine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant in the synthesis chiral 4, 4′-Bipyridines. 3-bromo-5-(trifluoromethyl)pyridine is prepared from 3-Bromo-5-iodopyridine.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light, heat, air.
TRUSTED_SUSTAINABILITY
Melting Point 127°C to 131°C
Color White
Quantity 250 mg
Beilstein 108478
Sensitivity Light sensitive
Synonym 3-bromo-5-iodo-pyridine, 5-bromo-3-iodopyridine, pyridine, 3-bromo-5-iodo, 3-bromo-5-iodpyridine, 3-iodo-5-bromopyridine, pubchem8080, acmc-1cdm9, pyridine,3-bromo-5-iodo, 3-bromo-5-iodopyridine, abbypharma ap-13-5787
Solubility Information Slightly soluble in water.
Formula Weight 283.89
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-Bromo-5-iodopyridine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'