missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-fluorobenzylamine hydrochloride, 96%, Thermo Scientific™

Catalog Number p-7054216
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
1189924-80-8
C7H8BrClFN
240.5
MFCD04113608
OFYIDRUSNSFKEG-UHFFFAOYSA-N
3-bromo-5-fluorophenyl methanamine hydrochloride, 3-bromo-5-fluorobenzylamine hydrochloride, 3-bromo-5-fluorophenyl methylamine, chloride, 3-bromanyl-5-fluoranyl-phenyl methanamine hydrochloride, 1-3-bromo-5-fluorophenyl methanamine hydrochloride
44717343
(3-bromo-5-fluorophenyl)methanamine;hydrochloride
C1=C(C=C(C=C1F)Br)CN.Cl
This item is not returnable. View return policy
1189924-80-8
C7H8BrClFN
240.5
MFCD04113608
OFYIDRUSNSFKEG-UHFFFAOYSA-N
3-bromo-5-fluorophenyl methanamine hydrochloride, 3-bromo-5-fluorobenzylamine hydrochloride, 3-bromo-5-fluorophenyl methylamine, chloride, 3-bromanyl-5-fluoranyl-phenyl methanamine hydrochloride, 1-3-bromo-5-fluorophenyl methanamine hydrochloride
44717343
(3-bromo-5-fluorophenyl)methanamine;hydrochloride
C1=C(C=C(C=C1F)Br)CN.Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 240.5
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 3-Bromo-5-fluorobenzylamine hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'