missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-fluorobenzyl alcohol, 98%, Thermo Scientific™

Catalog Number 15487948
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
216755-56-5
C7H6BrFO
205.026
MFCD04116320
KQWBQTOZXPPFIY-UHFFFAOYSA-N
3-bromo-5-fluorobenzyl alcohol, 3-bromo-5-fluorophenyl methanol, 3-bromo-5-fluorobenzylalcohol, 5-bromo-3-fluorobenzyl alcohol, benzenemethanol,3-bromo-5-fluoro, benzenemethanol, 3-bromo-5-fluoro, 3-bromo-5-fluorophenyl methan-1-ol, 3-bromo-5-fluoro-phenyl methanol, pubchem4885, acmc-1cgon
2769402
(3-bromo-5-fluorophenyl)methanol
C1=C(C=C(C=C1F)Br)CO
This item is not returnable. View return policy
216755-56-5
C7H6BrFO
205.026
MFCD04116320
KQWBQTOZXPPFIY-UHFFFAOYSA-N
3-bromo-5-fluorobenzyl alcohol, 3-bromo-5-fluorophenyl methanol, 3-bromo-5-fluorobenzylalcohol, 5-bromo-3-fluorobenzyl alcohol, benzenemethanol,3-bromo-5-fluoro, benzenemethanol, 3-bromo-5-fluoro, 3-bromo-5-fluorophenyl methan-1-ol, 3-bromo-5-fluoro-phenyl methanol, pubchem4885, acmc-1cgon
2769402
(3-bromo-5-fluorophenyl)methanol
C1=C(C=C(C=C1F)Br)CO

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color Colorless
Quantity 250 mg
Formula Weight 205.02
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Bromo-5-fluorobenzyl alcohol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'