missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-chlorobenzotrifluoride, 98%, Thermo Scientific™

Catalog Number p-7041380
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
928783-85-1
C7H3BrClF3
259.45
MFCD09260894
QRPRXRQFNNXPBA-UHFFFAOYSA-N
3-bromo-5-chlorobenzotrifluoride, 1-bromo-3-chloro-5-trifluoromethyl benzene, benzene, 1-bromo-3-chloro-5-trifluoromethyl, 5-bromo-3-chlorobenzotrifluoride, 3-bromo-5-chloro-alpha,alpha,alpha-trifluorotoluene, acmc-209ri6, ksc912o0j, 3-bromo-5-chloro-trifluoromethylbenzene
20269855
1-bromo-3-chloro-5-(trifluoromethyl)benzene
C1=C(C=C(C=C1Cl)Br)C(F)(F)F
This item is not returnable. View return policy
928783-85-1
C7H3BrClF3
259.45
MFCD09260894
QRPRXRQFNNXPBA-UHFFFAOYSA-N
3-bromo-5-chlorobenzotrifluoride, 1-bromo-3-chloro-5-trifluoromethyl benzene, benzene, 1-bromo-3-chloro-5-trifluoromethyl, 5-bromo-3-chlorobenzotrifluoride, 3-bromo-5-chloro-alpha,alpha,alpha-trifluorotoluene, acmc-209ri6, ksc912o0j, 3-bromo-5-chloro-trifluoromethylbenzene
20269855
1-bromo-3-chloro-5-(trifluoromethyl)benzene
C1=C(C=C(C=C1Cl)Br)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 25 g
Formula Weight 259.45
Percent Purity 98%
Chemical Name or Material 3-Bromo-5-chlorobenzotrifluoride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'