missing translation for 'onlineSavingsMsg'
Få mere at vide

3-Bromo-4-chloro-7-fluoroquinoline, TRC

Artikelnummer. 30343031
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30343031 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30343031 Leverandør Toronto Research Chemicals Leverandørnr. TRCB874593500MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 3-Bromo-4-chloro-7-fluoroquinoline
CAS 1204810-31-0
Molekylær formel C9H4NFClBr
Mængde 500 mg
InChI formel InChI=1S/C9H4BrClFN/c10-7-4-13-8-3-5(12)1-2-6(8)9(7)11/h1-4H
SMIL Fc1ccc2c(Cl)c(Br)cnc2c1
IUPAC navn 3-bromo-4-chloro-7-fluoroquinoline
Molekylvægt (g/mol) 260.49
Formel vægt 258.92
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.