missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-methylbenzoic acid, 97%, Thermo Scientific™

Catalog Number p-3757071
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
76006-33-2
C8H7BrO2
215.05
MFCD00270097
BJGKVCKGUBYULR-UHFFFAOYSA-N
3-bromo-o-toluic acid, 2-methyl-3-bromobenzoic acid, 3-bromo-2-methyl-benzoic acid, benzoic acid, 3-bromo-2-methyl, pubchem4717, bromo-methylbenzoic acid, 2-bromo-6-carboxytoluene, 3-bromo-2-methybenzoic acid, 3-bromo-2-methylbenzoicacid, ksc377a4p
2735590
3-bromo-2-methylbenzoic acid
CC1=C(C=CC=C1Br)C(O)=O
This item is not returnable. View return policy
76006-33-2
C8H7BrO2
215.05
MFCD00270097
BJGKVCKGUBYULR-UHFFFAOYSA-N
3-bromo-o-toluic acid, 2-methyl-3-bromobenzoic acid, 3-bromo-2-methyl-benzoic acid, benzoic acid, 3-bromo-2-methyl, pubchem4717, bromo-methylbenzoic acid, 2-bromo-6-carboxytoluene, 3-bromo-2-methybenzoic acid, 3-bromo-2-methylbenzoicacid, ksc377a4p
2735590
3-bromo-2-methylbenzoic acid
CC1=C(C=CC=C1Br)C(O)=O
CAS Min % 96.0
CAS Max % 100.0
Melting Point 152.0°C to 156.0°C
Color Brown or Orange to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 215.05
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3-Bromo-2-methylbenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 3-Bromo-2-methylbenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'