missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-2-methoxypyridine, 96%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Specifications
CAS | 13472-59-8 |
Density | 1.5860g/mL |
Boiling Point | 70°C to 75°C (3.0 Torr) |
Flash Point | 93°C |
Assay Percent Range | 95% min. (GC) |
Packaging | Glass Bottle |
Molecular Formula | C6H6BrNO |
MDL Number | MFCD03095349 |
Quantity | 1 g |
Specific Gravity | 1.586 |
Show More |
Safety and Handling
Product Identifier
- 3-Bromo-2-methoxypyridine
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'