missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-2-fluorobenzyl bromide, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
Used to obtain the target inhibitors. A new quasi-one-dimensional molecular solid based on Ni (mnt) 2 anion: Crystal structure and spin-gap transition 4-Bromo-2-fluorobenzyl bromide acts as stasrting material.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used to obtain the target inhibitors. A new quasi-one-dimensional molecular solid based on Ni (mnt) 2 anion: Crystal structure and spin-gap transition 4-Bromo-2-fluorobenzyl bromide acts as stasrting material.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Used to obtain the target inhibitors. A new quasi-one-dimensional molecular solid based on Ni (mnt) 2 anion: Crystal structure and spin-gap transition 4-Bromo-2-fluorobenzyl bromide acts as stasrting material.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Boiling Point | 60°C (0.1 mmHg) |
Quantity | 250 mg |
UN Number | UN3265 |
Synonym | 1-bromo-3-bromomethyl-2-fluorobenzene, 3-bromo-2-fluorobenzyl bromide, benzene,1-bromo-3-bromomethyl-2-fluoro, acmc-20ahso, intermediates-zcf02146, benzene, 1-bromo-3-bromomethyl-2-fluoro |
Formula Weight | 267.92 |
Percent Purity | 97% |
Chemical Name or Material | 3-Bromo-2-fluorobenzyl bromide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'