missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-fluoro-5-(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals

Catalog Number p-7056012
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
1031929-01-7
C6H2BrF4N
243.99
MFCD08061304
HKACHYLUXHBYMO-UHFFFAOYSA-N
3-bromo-2-fluoro-5-trifluoromethyl pyridine, 2-fluoro-3-bromo-5-trifluoromethylpyridine, pyridine, 3-bromo-2-fluoro-5-trifluoromethyl, 3-bromo-2-fluoro-5-trifluoromethyl ryridine, pubchem15149, pyridine,3-bromo-2-fluoro-5-trifluoromethyl
23053237
3-bromo-2-fluoro-5-(trifluoromethyl)pyridine
FC1=C(Br)C=C(C=N1)C(F)(F)F
This item is not returnable. View return policy
1031929-01-7
C6H2BrF4N
243.99
MFCD08061304
HKACHYLUXHBYMO-UHFFFAOYSA-N
3-bromo-2-fluoro-5-trifluoromethyl pyridine, 2-fluoro-3-bromo-5-trifluoromethylpyridine, pyridine, 3-bromo-2-fluoro-5-trifluoromethyl, 3-bromo-2-fluoro-5-trifluoromethyl ryridine, pubchem15149, pyridine,3-bromo-2-fluoro-5-trifluoromethyl
23053237
3-bromo-2-fluoro-5-(trifluoromethyl)pyridine
FC1=C(Br)C=C(C=N1)C(F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.774 g/mL
Flash Point 30°C (86°F)
Refractive Index 1.46
Quantity 250 mg
UN Number UN1992
Formula Weight 243.99
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Bromo-2-fluoro-5-(trifluoromethyl)pyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'