Få mere at vide
3-Bromo-1,2-propanediol-d5, TRC
Artikelnummer. 30336709
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30336709 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30336709 Leverandør Toronto Research Chemicals Leverandørnr. TRCB6866571MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Bromo-1,2-propanediol-d5
CAS 1246820-48-3
Renhedsgrad noter GC
Molekylær formel C3D5H2BrO2
Mængde 1 mg
Synonym (+/-)-Glycerol 1-Bromohydrin-d5, 1-Bromo-1-deoxyglycerol-d5, 1-Bromo-2,3-dihydroxy-propane-d5, 1-Bromo-2,3-propanediol-d5, 3-Bromo-1,2-dihydroxypropane-d5, DL-Glycerol 1-Bromohydrin-d5, Glycerol Bromohydrin-d5, Glycerol α-Bromohydrin-d5, α-Bromohydrin-d5
InChI formel InChI=1S/C3H7BrO2/c4-1-3(6)2-5/h3,5-6H,1-2H2/i1D2,2D2,3D
SMIL [2H]C([2H])(O)C([2H])(O)C([2H])([2H])Br
IUPAC navn 3-bromo-1,1,2,3,3-pentadeuterio-propane-1,2-diol
Molekylvægt (g/mol) 160.02
Formel vægt 158.9943
Procent renhed >95