missing translation for 'onlineSavingsMsg'
Learn More

3-(BOC-Amino)piperidine, 97%, Thermo Scientific Chemicals

Catalog Number p-4228401
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
172603-05-3
C10H20N2O2
200.28
WUOQXNWMYLFAHT-UHFFFAOYSA-N
3-boc-aminopiperidine, tert-butyl piperidin-3-ylcarbamate, 3-n-boc-aminopiperidine, tert-butyl n-piperidin-3-yl carbamate, 3-boc-amino piperidine, 3-n-boc-amino piperidine, 3-tert-butoxycarbonylamino piperidine, 3-aminopiperidine, 3-boc protected, piperidin-3-yl-carbamic acid tert-butyl ester, tert-butyl n-3-piperidyl carbamate
4233041
tert-butyl N-piperidin-3-ylcarbamate
CC(C)(C)OC(=O)NC1CCCNC1
This item is not returnable. View return policy
172603-05-3
C10H20N2O2
200.28
WUOQXNWMYLFAHT-UHFFFAOYSA-N
3-boc-aminopiperidine, tert-butyl piperidin-3-ylcarbamate, 3-n-boc-aminopiperidine, tert-butyl n-piperidin-3-yl carbamate, 3-boc-amino piperidine, 3-n-boc-amino piperidine, 3-tert-butoxycarbonylamino piperidine, 3-aminopiperidine, 3-boc protected, piperidin-3-yl-carbamic acid tert-butyl ester, tert-butyl n-3-piperidyl carbamate
4233041
tert-butyl N-piperidin-3-ylcarbamate
CC(C)(C)OC(=O)NC1CCCNC1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 104.6°C to 112.6°C
CAS Min % 96.0
CAS Max % 100.0
Color White
Assay Percent Range 96% min. (GC)
Infrared Spectrum Authentic
Packaging Glass Bottle
Formula Weight 200.28
Percent Purity 97%
Quantity 1 g
Physical Form Crystalline Powder
Chemical Name or Material 3-(BOC-Amino)piperidine
Show More Show Less
compliance-icons
Product Identifier
  • 3-(BOC-Amino)piperidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'