missing translation for 'onlineSavingsMsg'
Learn More

3-(Boc-amino)-3-(4-chlorophenyl)propionic acid, 97%, Thermo Scientific™

Catalog Number p-7047821
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
284493-65-8
C14H17ClNO4
298.74
MFCD02090708,MFCD03427960
ZPXVKCUGZBGIBW-NSHDSACASA-M
3-boc-amino-3-4-chlorophenyl propionic acid, 3-tert-butoxycarbonyl amino-3-4-chlorophenyl propanoic acid, 3-boc-amino-3-4'-chlorophenyl propioinic acid, 3-tert-butoxycarbonylamino-3-4-chloro-phenyl-propionic acid, 3-n-boc-amino-3-4-chlorophenyl propionic acid, 3-tert-butoxycarbonylamino-3-4-chlorophenyl propanoic acid, 3-tert-butoxy carbonylamino-3-4-chlorophenyl propanoic acid, boc-r-3-amino-3-4-chloro-phenyl-propionic acid
2734402
3-(4-chlorophenyl)-3-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)N[C@@H](CC([O-])=O)C1=CC=C(Cl)C=C1
This item is not returnable. View return policy
284493-65-8
C14H17ClNO4
298.74
MFCD02090708,MFCD03427960
ZPXVKCUGZBGIBW-NSHDSACASA-M
3-boc-amino-3-4-chlorophenyl propionic acid, 3-tert-butoxycarbonyl amino-3-4-chlorophenyl propanoic acid, 3-boc-amino-3-4'-chlorophenyl propioinic acid, 3-tert-butoxycarbonylamino-3-4-chloro-phenyl-propionic acid, 3-n-boc-amino-3-4-chlorophenyl propionic acid, 3-tert-butoxycarbonylamino-3-4-chlorophenyl propanoic acid, 3-tert-butoxy carbonylamino-3-4-chlorophenyl propanoic acid, boc-r-3-amino-3-4-chloro-phenyl-propionic acid
2734402
3-(4-chlorophenyl)-3-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
CC(C)(C)OC(=O)N[C@@H](CC([O-])=O)C1=CC=C(Cl)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 179°C to 180°C (decomposition)
Quantity 250 mg
Formula Weight 299.75
Percent Purity 97%
Chemical Name or Material 3-(Boc-amino)-3-(4-chlorophenyl)propionic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'