Få mere at vide
3-Benzyloxy-Alpha-(N-butyryl-d3)-aminopropionitrile, TRC
Artikelnummer. 30336736
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30336736 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30336736 Leverandør Toronto Research Chemicals Leverandørnr. TRCB25885250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Benzyloxy-alpha-(N-butyryl-d3)-aminopropionitrile
CAS 1216615-94-9
Molekylær formel C15H17D3N2O2
Mængde 50 mg
Synonym N-[1-Cyano-2-(phenylmethoxy)ethyl]pentanamide-d3
InChI formel InChI=1S/C15H20N2O2/c1-2-3-9-15(18)17-14(10-16)12-19-11-13-7-5-4-6-8-13/h4-8,14H,2-3,9,11-12H2,1H3,(H,17,18)/i1D3
SMIL [2H]C([2H])([2H])CCCC(=O)NC(COCc1ccccc1)C#N
IUPAC navn N-(1-cyano-2-phenylmethoxyethyl)-5,5,5-trideuteriopentanamide
Molekylvægt (g/mol) 263.35
Formel vægt 263.1713