missing translation for 'onlineSavingsMsg'
Learn More

3-Benzyl-5-chloromethyl-1,2,4-oxadiazole, 97%, Thermo Scientific™

Catalog Number p-7039234
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
51802-77-8
C10H9ClN2O
208.645
MFCD08273449
RUERNNLQCZMFKW-UHFFFAOYSA-N
3-benzyl-5-chloromethyl-1,2,4-oxadiazole, 1,2,4-oxadiazole, 5-chloromethyl-3-phenylmethyl, 5-chloromethyl-3-benzyl-1,2,4-oxadiazole
8027132
3-benzyl-5-(chloromethyl)-1,2,4-oxadiazole
C1=CC=C(C=C1)CC2=NOC(=N2)CCl
This item is not returnable. View return policy
51802-77-8
C10H9ClN2O
208.645
MFCD08273449
RUERNNLQCZMFKW-UHFFFAOYSA-N
3-benzyl-5-chloromethyl-1,2,4-oxadiazole, 1,2,4-oxadiazole, 5-chloromethyl-3-phenylmethyl, 5-chloromethyl-3-benzyl-1,2,4-oxadiazole
8027132
3-benzyl-5-(chloromethyl)-1,2,4-oxadiazole
C1=CC=C(C=C1)CC2=NOC(=N2)CCl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 2 g
Formula Weight 208.65
Percent Purity 97%
Chemical Name or Material 3-Benzyl-5-chloromethyl-1,2,4-oxadiazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'