missing translation for 'onlineSavingsMsg'
Få mere at vide

3-Aminopropionitrile Fumarate (2:1), TRC

Artikelnummer. 30319360
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30319360 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30319360 Leverandør Toronto Research Chemicals Leverandørnr. TRCA628190100G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 3-Aminopropionitrile Fumarate (2:1)
CAS 2079-89-2
Molekylær formel C10H16N4O4
Mængde 100 g
Synonym 3-Amino-propanenitrile (2E)-2-Butenedioate (2:1), Fumaric acid salt with 3-aminopropionitrile, 3-Amino-propanenitrile (E)-2-Butenedioate (2:1), 3-Amino-propionitrile Fumarate (2:1), 3-Aminopropionitrile fumarate (2:1), Di-beta-aminopropionitrile fumarate, beta-Aminopropionitrile fumarate, beta-Ammoniumpropionitrile hemifumarate
InChI formel InChI=1S/C4H4O4.2C3H6N2/c5-3(6)1-2-4(7)8;2*4-2-1-3-5/h1-2H,(H,5,6)(H,7,8);2*1-2,4H2/b2-1+;;
SMIL NCCC#N.NCCC#N.OC(=O)\C=C\C(=O)O
IUPAC navn 3-aminopropanenitrile;(E)-but-2-enedioic acid
Molekylvægt (g/mol) 256.26
Formel vægt 256.1172
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.