missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
(3-Aminophenyl)acetic Acid, TRC
Specifications
Specifications
| Kemisk navn eller materiale | (3-Aminophenyl)acetic Acid |
| CAS | 14338-36-4 |
| Molekylær formel | C8 H9 N O2 |
| Mængde | 500 mg |
| Synonym | Benzeneacetic acid, 3-amino-, Acetic acid, (m-aminophenyl)- (6CI,7CI), 3-Aminobenzeneacetic acid, (3-Aminophenyl)acetic acid, (m-Aminophenyl)acetic acid, 2-(3-Aminophenyl)acetic acid |
| InChI formel | InChI=1S/C8H9NO2/c9-7-3-1-2-6(4-7)5-8(10)11/h1-4H,5,9H2,(H,10,11) |
| SMIL | Nc1cccc(CC(=O)O)c1 |
| IUPAC navn | 2-(3-aminophenyl)acetic acid |
| Molekylvægt (g/mol) | 151.16 |
| Formel vægt | 151.0633 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?