Få mere at vide
3-Aminopentane-d5, TRC
Artikelnummer. 30322330
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30322330 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322330 Leverandør Toronto Research Chemicals Leverandørnr. TRCA61895210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Aminopentane-2,2,3,4,4-D5
CAS 1219802-43-3
Molekylær formel C5 D11 H2 N
Mængde 10 mg
Synonym 2,2,3,4,4-Pentadeuteriopentan-3-amine, 3-Pentanamine-2,2,3,4,4-d5, 1-(Ethyl-1,1-d2)propylamine-1,2,2-d3, 3-Aminopentane-2,2,3,4,4-d5, 3-Aminopentane-2,2,3,4,4-D5, 3-Pentylamine-2,2,3,4,4-d5
InChI formel InChI=1S/C5H13N/c1-3-5(6)4-2/h5H,3-4,6H2,1-2H3/i1D3,2D3,3D2,4D2,5D
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])(N)C([2H])([2H])C([2H])([2H])[2H]
IUPAC navn 1,1,1,2,2,3,4,4,5,5,5-undecadeuteriopentan-3-amine
Molekylvægt (g/mol) 98.2312
Formel vægt 98.1738