missing translation for 'onlineSavingsMsg'
Learn More

3-Aminomethyltetrahydrofuran, 97%, Thermo Scientific Chemicals

Catalog Number p-4229065
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
165253-31-6
C5H11NO
101.15
MFCD08234925
CINJIXGRSTYIHP-UHFFFAOYSA-N
tetrahydrofuran-3-yl methanamine, tetrahydro-3-furanmethanamine, tetrahydrofuran-3-ylmethylamine, 3-furanmethanamine, tetrahydro, 1-tetrahydrofuran-3-ylmethanamine, c-tetrahydro-furan-3-yl-methylamine, 3-aminomethyl oxolane, 3-aminomethyl tetrahydrofuran, tetrahydrofur-3-yl methylamine, c-tetrahydrofuran-3-yl methylamine
10898660
oxolan-3-ylmethanamine
C1COCC1CN
This item is not returnable. View return policy
165253-31-6
C5H11NO
101.15
MFCD08234925
CINJIXGRSTYIHP-UHFFFAOYSA-N
tetrahydrofuran-3-yl methanamine, tetrahydro-3-furanmethanamine, tetrahydrofuran-3-ylmethylamine, 3-furanmethanamine, tetrahydro, 1-tetrahydrofuran-3-ylmethanamine, c-tetrahydro-furan-3-yl-methylamine, 3-aminomethyl oxolane, 3-aminomethyl tetrahydrofuran, tetrahydrofur-3-yl methylamine, c-tetrahydrofuran-3-yl methylamine
10898660
oxolan-3-ylmethanamine
C1COCC1CN

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Boiling Point 65.0°C (15.0 Torr)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
UN Number 2924
Formula Weight 101.15
Percent Purity 97%
Physical Form Solution
Chemical Name or Material 3-Aminomethyltetrahydrofuran
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Aminomethyltetrahydrofuran
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'