Få mere at vide
3-[(Aminocarbonyl)amino]-5-[4-(4-morpholinylmethyl)phenyl]-2-thiophenecarboxamide, TRC
Artikelnummer. 30339625
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30339625 5 mg
30337163 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30339625 Leverandør Toronto Research Chemicals Leverandørnr. TRCA7948005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-[(Aminocarbonyl)amino]-5-[4-(4-morpholinylmethyl)phenyl]-2-thiophenecarboxamide
CAS 494772-86-0
Molekylær formel C17H20N4O3S
Mængde 5 mg
Synonym 3-[(Aminocarbonyl)amino]-5-[4-(morpholin-4-ylmethyl)phenyl]thiophene-2-carboxamide
InChI formel InChI=1S/C17H20N4O3S/c18-16(22)15-13(20-17(19)23)9-14(25-15)12-3-1-11(2-4-12)10-21-5-7-24-8-6-21/h1-4,9H,5-8,10H2,(H2,18,22)(H3,19,20,23)
SMIL O=C(N)C(S1)=C(NC(N)=O)C=C1C2=CC=C(C=C2)CN3CCOCC3
IUPAC navn 3-(carbamoylamino)-5-[4-(morpholin-4-ylmethyl)phenyl]thiophene-2-carboxamide
Molekylvægt (g/mol) 360.43
Formel vægt 360.1256