missing translation for 'onlineSavingsMsg'
Få mere at vide

3-Aminobenzoic-d4 Acid, TRC

Artikelnummer. 30311714
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30311714 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30311714 Leverandør Toronto Research Chemicals Leverandørnr. TRCA591492100MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 3-Aminobenzoic-d4 Acid
CAS 78399-79-8
Molekylær formel C7 D4 H3 N O2
Mængde 100 mg
Synonym Benzoic-2,3,4,6-d4 acid, 5-amino- (9CI), 3-Aminobenzoic-2,4,5,6-d4 Acid, 3-Aminobenzoic Acid-2,4,5,6-D4, 3-Aminobenzoic Acid-d4
InChI formel InChI=1S/C7H7NO2/c8-6-3-1-2-5(4-6)7(9)10/h1-4H,8H2,(H,9,10)/i1D,2D,3D,4D
SMIL [2H]c1c([2H])c(N)c([2H])c(C(=O)O)c1[2H]
IUPAC navn 3-amino-2,4,5,6-tetradeuteriobenzoic acid
Molekylvægt (g/mol) 141.1606
Formel vægt 141.0728
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.