missing translation for 'onlineSavingsMsg'
Få mere at vide

3-Aminobenzoic-d4 Acid Methyl Ester, TRC

Artikelnummer. 30326677
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30326677 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30326677 Leverandør Toronto Research Chemicals Leverandørnr. TRCA59149710MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 3-Aminobenzoic acid ring-d4 methyl ester
CAS 911132-57-5
Molekylær formel C8 2H4 H5 N O2
Mængde 10 mg
Synonym Benzoic-2,3,4,6-d4 acid, 5-amino-, methyl ester (9CI), 3-(Methoxycarbonyl)aniline-2,4,5,6-d4, 3-Aminobenzoic-2,4,5,6-d4 acid methyl ester, 3-Aminobenzoic-d4 acid methyl ester, 3-Aminobenzoic acid-d4 methyl ester, 3-Carbomethoxyaniline-2,4,5,6-d4, Methyl 3-aminobenzoate-d4, Methyl 3-aminobenzoate-2,4,5,6-D4, Methyl m-aminobenzoate-d4, m-Aminobenzoic acid-d4 methyl ester
InChI formel InChI=1S/C8H9NO2/c1-11-8(10)6-3-2-4-7(9)5-6/h2-5H,9H2,1H3/i2D,3D,4D,5D
SMIL [2H]c1c([2H])c(N)c([2H])c(C(=O)OC)c1[2H]
IUPAC navn methyl 3-amino-2,4,5,6-tetradeuteriobenzoate
Molekylvægt (g/mol) 155.19
Formel vægt 155.0884
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.