Få mere at vide
3-Amino-6-bromo-4-(2-chlorophenyl)-2(1H)-quinolinone-d5, TRC
Artikelnummer. 30310548
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30310548 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30310548 Leverandør Toronto Research Chemicals Leverandørnr. TRCA60195210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Amino-6-bromo-4-(2-chlorophenyl)-2(1H)-quinolinone-d5
CAS 1794793-32-0
Molekylær formel C15H5D5BrClN2O
Mængde 10 mg
InChI formel InChI=1S/C15H10BrClN2O/c16-8-5-6-12-10(7-8)13(14(18)15(20)19-12)9-3-1-2-4-11(9)17/h1-7H,18H2,(H,19,20)/i1D,2D,3D,4D,6D
SMIL [2H]c1cc(Br)cc2C(=C(N)C(=O)Nc12)c3c([2H])c([2H])c([2H])c([2H])c3Cl
IUPAC navn 3-amino-6-bromo-4-(2-chloro-3,4,5,6-tetradeuteriophenyl)-8-deuterio-1H-quinolin-2-one
Molekylvægt (g/mol) 354.64
Formel vægt 352.9979