missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-bromo-1H-indazole, 97%, Thermo Scientific Chemicals

Catalog Number 15436167
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
61272-71-7
C7H6BrN3
212.05
MFCD03426696
OMPYFDJVSAMSMA-UHFFFAOYSA-N
3-amino-5-bromo-1h-indazole, 3-amino-5-bromoindazole, 5-bromo-1h-indazol-3-ylamine, 1h-indazol-3-amine, 5-bromo, 5-bromo-1h-indazole-3-ylamine, pubchem18573, 3-amino-5-bromo-indazole, acmc-1b3h9, 5-bromo-1h-indazole-3-amine
817910
5-bromo-1H-indazol-3-amine
NC1=NNC2=CC=C(Br)C=C12
This item is not returnable. View return policy
61272-71-7
C7H6BrN3
212.05
MFCD03426696
OMPYFDJVSAMSMA-UHFFFAOYSA-N
3-amino-5-bromo-1h-indazole, 3-amino-5-bromoindazole, 5-bromo-1h-indazol-3-ylamine, 1h-indazol-3-amine, 5-bromo, 5-bromo-1h-indazole-3-ylamine, pubchem18573, 3-amino-5-bromo-indazole, acmc-1b3h9, 5-bromo-1h-indazole-3-amine
817910
5-bromo-1H-indazol-3-amine
NC1=NNC2=CC=C(Br)C=C12

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 171°C to 174°C
Quantity 5 g
Formula Weight 212.05
Percent Purity 97%
Chemical Name or Material 3-Amino-5-bromo-1H-indazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'