missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-methylphenylboronic acid, 98%, Thermo Scientific™

Catalog Number p-7077533
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
22237-12-3
C7H11BClNO2
187.43
MFCD01632201
BKFDOPYYSBQWIK-UHFFFAOYSA-N
3-amino-4-methylphenyl boronic acid, 3-amino-4-methylbenzeneboronic acid, 3-amino-4-methyl-phenyl boronic acid, 3-amino-p-tolylboronic acid, boronic acid, 3-amino-4-methylphenyl, acmc-20agxu, pubchem1745, 3-amino-4-methylphenylboranic acid, 3-amino4methyl phenyl boronic acid
2737803
Cl.CC1=CC=C(C=C1N)B(O)O
This item is not returnable. View return policy
22237-12-3
C7H11BClNO2
187.43
MFCD01632201
BKFDOPYYSBQWIK-UHFFFAOYSA-N
3-amino-4-methylphenyl boronic acid, 3-amino-4-methylbenzeneboronic acid, 3-amino-4-methyl-phenyl boronic acid, 3-amino-p-tolylboronic acid, boronic acid, 3-amino-4-methylphenyl, acmc-20agxu, pubchem1745, 3-amino-4-methylphenylboranic acid, 3-amino4methyl phenyl boronic acid
2737803
Cl.CC1=CC=C(C=C1N)B(O)O
Melting Point 250.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
IUPAC Name (3-amino-4-methylphenyl)boronic acid hydrochloride
Formula Weight 150.98
Percent Purity 98%
Chemical Name or Material 3-Amino-4-methylphenylboronic acid
compliance-icons
Product Identifier
  • 3-Amino-4-methylphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'