missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Amino-4-bromo-1H-indazole, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Microwave heating of 2-halobenzaldehydes or 2-haloacetophenones with phenylhydrazines at 160°C for 10 min quantitatively yielded the arylhydrazones, which were further cyclized to give 1-aryl-1H-indazoles via CuI/diamine-catalyzed N-arylation under microwave heating (160°C, 10 min). Good to excellent yields were observed for 2-iodo, 2-bromo, and 2-chloro benzaldehydes or acetophenones.
Solubility
Slightly soluble.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Refrigerate 2-7°C.
Microwave heating of 2-halobenzaldehydes or 2-haloacetophenones with phenylhydrazines at 160°C for 10 min quantitatively yielded the arylhydrazones, which were further cyclized to give 1-aryl-1H-indazoles via CuI/diamine-catalyzed N-arylation under microwave heating (160°C, 10 min). Good to excellent yields were observed for 2-iodo, 2-bromo, and 2-chloro benzaldehydes or acetophenones.
Solubility
Slightly soluble.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Refrigerate 2-7°C.
Specifications
Melting Point | 142°C to 144°C |
Quantity | 1 g |
Solubility Information | Slightly soluble. |
Formula Weight | 212.05 |
Percent Purity | 97% |
Chemical Name or Material | 3-Amino-4-bromo-1H-indazole |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'