missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4,6-dimethylisoxazolo[5,4-b]pyridine, 95%, Thermo Scientific™

Catalog Number p-7033110
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
916792-12-6
C8H9N3O
163.18
MFCD08060940
DYMSNXTUZQZFSM-UHFFFAOYSA-N
4,6-dimethylisoxazolo 5,4-b pyridin-3-ylamine, 4,6-dimethylisoxazolo 5,4-b pyridin-3-amine, 3-amino-4,6-dimethylisoxazolo 5,4-b pyridine, 4,6-dimethyl-1,2 oxazolo 5,4-b pyridin-3-amine, 4,6-dimethyl 1,2 oxazolo 5,4-b pyridin-3-amine, 4,6-dimethylisoxazolo 5,4-b pyridine-3-ylamine, acmc-20ana9
24208795
4,6-dimethyl-[1,2]oxazolo[5,4-b]pyridin-3-amine
CC1=CC(C)=C2C(N)=NOC2=N1
This item is not returnable. View return policy
916792-12-6
C8H9N3O
163.18
MFCD08060940
DYMSNXTUZQZFSM-UHFFFAOYSA-N
4,6-dimethylisoxazolo 5,4-b pyridin-3-ylamine, 4,6-dimethylisoxazolo 5,4-b pyridin-3-amine, 3-amino-4,6-dimethylisoxazolo 5,4-b pyridine, 4,6-dimethyl-1,2 oxazolo 5,4-b pyridin-3-amine, 4,6-dimethyl 1,2 oxazolo 5,4-b pyridin-3-amine, 4,6-dimethylisoxazolo 5,4-b pyridine-3-ylamine, acmc-20ana9
24208795
4,6-dimethyl-[1,2]oxazolo[5,4-b]pyridin-3-amine
CC1=CC(C)=C2C(N)=NOC2=N1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 163.18
Percent Purity 95%
Chemical Name or Material 3-Amino-4,6-dimethylisoxazolo[5,4-b]pyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'