Få mere at vide
3-Amino-2-pyrazinecarboxylic Acid Methyl Ester, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 3-Amino-2-pyrazinecarboxylic Acid Methyl Ester |
| CAS | 16298-03-6 |
| Molekylær formel | C6 H7 N3 O2 |
| Mængde | 1 g |
| Synonym | 2-Pyrazinecarboxylic acid, 3-amino-, methyl ester, Pyrazinecarboxylic acid, 3-amino-, methyl ester (7CI,8CI,9CI), Pyrazinoic acid, 3-amino-, methyl ester (6CI), 2-Amino-3-(methoxycarbonyl)pyrazine, 3-Aminopyrazine-2-carboxylic acid methyl ester, Methyl 2-aminopyrazine-3-carboxylate, Methyl 3-aminopyrazine-2-carboxylate, Methyl 3-aminopyrazinecarboxylate, NSC 123649 |
| InChI formel | InChI=1S/C6H7N3O2/c1-11-6(10)4-5(7)9-3-2-8-4/h2-3H,1H3,(H2,7,9) |
| SMIL | COC(=O)c1nccnc1N |
| IUPAC navn | methyl 3-aminopyrazine-2-carboxylate |
| Molekylvægt (g/mol) | 153.14 |
| Formel vægt | 153.0538 |