Learn More
3-Amino-2-naphthol, TRC
Product Code. 30348575
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity Unit Size
30348575 100 mg 100mg
1 options
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Product Code. 30348575 Supplier Toronto Research Chemicals Supplier No. TRCB404810100MG
 Restricted Item
Forventet forsendelse: 10-12-2026
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Amino-2-naphthol
CAS 5417-63-0
Molecular Formula C10 H9 N O
Quantity 100 mg
Synonym 2-Naphthol, 3-amino- (7CI, 8CI), 3-Amino-2-naphthalenol (ACI), 2-Amino-3-hydroxynaphthalene, 2-Amino-3-naphthol, 3-Amino-2-hydroxynaphthalene, 3-Amino-2-naphthol, 3-Amino-β-naphthol, 3-Hydroxy-2-aminonaphthalene, 3-Hydroxy-2-naphthylamine, NSC 7941, 2-Naphthalenol, 3-amino- (9CI, ACI)
InChI Formula InChI=1S/C10H9NO/c11-9-5-7-3-1-2-4-8(7)6-10(9)12/h1-6,12H,11H2
SMILES Nc1cc2ccccc2cc1O
IUPAC Name 3-aminonaphthalen-2-ol
Molecular Weight (g/mol) 159.184
Formula Weight 159.0684