Få mere at vide
3-Amino-2-methyl-benzyl-d2 Bromide Hydrobromide, TRC
Artikelnummer. 30314896
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30314896 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30314896 Leverandør Toronto Research Chemicals Leverandørnr. TRCA61437250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 3-Amino-2-methyl-benzyl-d2 Bromide Hydrobromide
CAS 57414-77-4
Molekylær formel C8 2H2 H8 Br N . Br H
Mængde 50 mg
InChI formel InChI=1S/C8H10BrN.BrH/c1-6-7(5-9)3-2-4-8(6)10;/h2-4H,5,10H2,1H3;1H/i5D2;
SMIL Br.[2H]C([2H])(Br)c1cccc(N)c1C
IUPAC navn 3-[bromo(dideuterio)methyl]-2-methylaniline;hydrobromide
Molekylvægt (g/mol) 283
Formel vægt 280.9384