missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Amino-2-methyl-6-(trifluoromethyl)pyridine, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed containers. With adequate ventilation. Store away from oxidizing agent.
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed containers. With adequate ventilation. Store away from oxidizing agent.
Specifications
Melting Point | 106°C to 108°C |
Quantity | 250 mg |
Synonym | 2-methyl-6-trifluoromethyl pyridin-3-amine, 2-methyl-6-trifluoromethyl-3-pyridinamine, 3-amino-2-methyl-6-trifluoromethyl pyridine, 2-methyl-6-trifluoromethylpyridin-3-yl amine, 2-methyl-6-trifluoromethyl-pyridin-3-ylamine, 2-methyl-6-trifluoromethyl-3-pyridylamine, 3-pyridinamine,2-methyl-6-trifluoromethyl, 2-methyl-6-trifluoromethyl pyridine-3-amine, 3-pyridinamine, 2-methyl-6-trifluoromethyl |
Solubility Information | Slightly soluble in water. |
Formula Weight | 176.14 |
Percent Purity | 97% |
Chemical Name or Material | 3-Amino-2-methyl-6-(trifluoromethyl)pyridine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'