missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2-methyl-6-(trifluoromethyl)pyridine, 97%, Thermo Scientific™

Catalog Number 15438545
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
383907-17-3
C7H7F3N2
176.14
MFCD01311991
FUJKUIAVFVGTGS-UHFFFAOYSA-N
3521850
2-methyl-6-(trifluoromethyl)pyridin-3-amine
CC1=C(N)C=CC(=N1)C(F)(F)F
This item is not returnable. View return policy
383907-17-3
C7H7F3N2
176.14
MFCD01311991
FUJKUIAVFVGTGS-UHFFFAOYSA-N
3521850
2-methyl-6-(trifluoromethyl)pyridin-3-amine
CC1=C(N)C=CC(=N1)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed containers. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Melting Point 106°C to 108°C
Quantity 250 mg
Synonym 2-methyl-6-trifluoromethyl pyridin-3-amine, 2-methyl-6-trifluoromethyl-3-pyridinamine, 3-amino-2-methyl-6-trifluoromethyl pyridine, 2-methyl-6-trifluoromethylpyridin-3-yl amine, 2-methyl-6-trifluoromethyl-pyridin-3-ylamine, 2-methyl-6-trifluoromethyl-3-pyridylamine, 3-pyridinamine,2-methyl-6-trifluoromethyl, 2-methyl-6-trifluoromethyl pyridine-3-amine, 3-pyridinamine, 2-methyl-6-trifluoromethyl
Solubility Information Slightly soluble in water.
Formula Weight 176.14
Percent Purity 97%
Chemical Name or Material 3-Amino-2-methyl-6-(trifluoromethyl)pyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'