Få mere at vide
3’-Amino-2’-Hydroxy-[1,1’]biphenyl-3-carboxylic Acid, TRC
Artikelnummer. 30136608
Skift visning
Mængde:
250 mg
2.5 g
Pakningsstørrelse:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30136608 2.5 g 2.5g
30012064 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30136608 Leverandør Toronto Research Chemicals Leverandørnr. TRCA6112552.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 376592-93-7
Molekylær formel C13 H11 N O3
Molekylvægt (g/mol) 229.231
Synonym [1,1'-Biphenyl]-3-carboxylic acid, 3'-amino-2'-hydroxy-, 3'-Amino-2'-hydroxy[1,1'-biphenyl]-3-carboxylic acid, 3'-Amino-2'-hydroxy-[1,1'-biphenyl]-3-carboxylic acid
IUPAC navn 3-(3-amino-2-hydroxyphenyl)benzoic acid
SMIL Nc1cccc(c1O)c2cccc(c2)C(=O)O
Tekniske data
Kemisk navn eller materiale 3-(3-amino-2-hydroxyphenyl)benzoic acid
Mængde 2.5 g
InChI formel InChI=1S/C13H11NO3/c14-11-6-2-5-10(12(11)15)8-3-1-4-9(7-8)13(16)17/h1-7,15H,14H2,(H,16,17)
Formel vægt 229.074