Få mere at vide
3-Amino-2-chloro-4-methylpyridine, 97%, Thermo Scientific™
Artikelnummer. 11381989
Skift visning
Mængde:
1 g
5 g
Pakningsstørrelse:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
11381989 1 g 1g
11391989 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 11381989 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. H26058.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 133627-45-9
Molekylær formel C6H7ClN2
Molekylvægt (g/mol) 142.59
MDL nummer MFCD00673152
InChI nøgle UOBCYTOUXLAABU-UHFFFAOYSA-N
Synonym 3-amino-2-chloro-4-methylpyridine, 2-chloro-3-amino-4-methylpyridine, 3-amino-2-chloro-4-picoline, 2-chloro-3-amino-4-methyl pyridine, 2-chloro-4-methyl-pyridin-3-ylamine, 2-chloro-3-amino-4-picoline, 2-chloro-4-methyl-3-pyridinamine, 3-pyridinamine, 2-chloro-4-methyl, 2-chloro-4-methyl-3-pyridylamine, 3-amino-2-chloro-4-methyl pyridine
PubChem CID 2756387
IUPAC navn 2-chloro-4-methylpyridin-3-amine
SMIL CC1=CC=NC(Cl)=C1N
Tekniske data
Kemisk navn eller materiale 3-Amino-2-chloro-4-methylpyridine
Smeltepunkt 64°C to 68°C
Mængde 1 g
Formel vægt 142.59
Procent renhed 97%