missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2-bromo-6-methylpyridine, 98%, Thermo Scientific Chemicals

Catalog Number p-7055663
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
126325-53-9
C6H7BrN2
187.04
MFCD16606103
ORELOTUWTIKRIW-UHFFFAOYSA-N
3-amino-4-bromo-2-methylpyridine, ksc915a1f, 3-pyridinamine, 4-bromo-2-methyl, 4-bromo-2-methyl-3-pyridinamine
14418044
4-bromo-2-methylpyridin-3-amine
CC1=NC=CC(Br)=C1N
This item is not returnable. View return policy
126325-53-9
C6H7BrN2
187.04
MFCD16606103
ORELOTUWTIKRIW-UHFFFAOYSA-N
3-amino-4-bromo-2-methylpyridine, ksc915a1f, 3-pyridinamine, 4-bromo-2-methyl, 4-bromo-2-methyl-3-pyridinamine
14418044
4-bromo-2-methylpyridin-3-amine
CC1=NC=CC(Br)=C1N

It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Color White
Quantity 250 mg
UN Number UN2811
Beilstein 4243986
Solubility Information Slightly soluble in water.
Formula Weight 187.04
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3-Amino-2-bromo-6-methylpyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'